2-Cyanphenylacetonitril

Lieferant: FLUOROCHEM

021667-100G 021667-25G
FLUO021667-100GEA 317 EUR
FLUO021667-100G FLUO021667-25G
2-Cyanphenylacetonitril
2-Cyanphenylacetonitril
Formel: C₉H₆N₂
MDL: MFCD00001895
CAS-Nummer: 3759-28-2
EINECS: 223-167-3
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR