3-(2-Chlorphenyl)-5-methyl-4-isoxazolcarbonsäure

Lieferant: FLUOROCHEM

021686-100MG
FLUO021686-100MGEA 0 EUR
FLUO021686-100MG
3-(2-Chlorphenyl)-5-methyl-4-isoxazolcarbonsäure
3-(2-Chlorophenyl)-5-methylisoxazole-4-carboxylic acid
Formel: C₁₁H₈ClNO₃
Molecular Weight: 237,64 g/mol
MDL: MFCD00020813
CAS-Nummer: 23598-72-3
EINECS: 245-770-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR