3-Fluor-4-methoxyphenylboronsäure

Lieferant: FLUOROCHEM

Synonyme: (3-Fluoro-4-methoxyphenyl)boronic acid, 3-Fluoro-4-methoxybenzeneboronic acid

021710-5G
FLUO021710-5GEA 36.9 EUR
FLUO021710-5G
3-Fluor-4-methoxyphenylboronsäure
3-Fluor-4-methoxyphenylboronsäure
Formel: C₇H₈BFO₃
MDL: MFCD00807404
CAS-Nummer: 149507-26-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR