4-Acetylbenzoesäure

Lieferant: FLUOROCHEM

Synonyme: Acetophenon-4-carbonsäure

021711-5G 021711-25G
FLUO021711-5GEA 39.9 EUR
FLUO021711-5G FLUO021711-25G
4-Acetylbenzoesäure
4-Acetylbenzoesäure
Formel: C₉H₈O₃
Molecular Weight: 164.16 g/mol
Siedepunkt: 340
MDL: MFCD00002561
CAS-Nummer: 586-89-0
EINECS: 209-588-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR