2-Bromthiazol
Lieferant: FLUOROCHEM
021802-25G
FLUO021802-25GEA
45.9
EUR
FLUO021802-25G
2-Bromthiazol
2-Bromothiazole
Formel:
C₃H₂BrNS Siedepunkt: 171 °C (1013 hPa) Dichte: 1,82 g/cm³ (20 °C) |
MDL:
MFCD00005316 CAS-Nummer: 3034-53-5 EINECS: 221-229-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...