6-Fluor-m-toluidin
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
3-Amino-4-fluortoluol, 2-Fluor-5-methylanilin
021823-50G
021823-10G
FLUO021823-50GEA
200
EUR
FLUO021823-50G
FLUO021823-10G
6-Fluor-m-toluidin
6-Fluor-m-toluidin
Formel:
C₇H₈FN Molecular Weight: 125.15 g/mol Siedepunkt: 88…90 °C (17 mmHg) Dichte: 1.109 g/cm³ (20 °C) |
MDL:
MFCD00007654 CAS-Nummer: 452-84-6 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...