3,4-Dimethoxyphenylboronsäurepinakolester

Lieferant: FLUOROCHEM

Synonyme: 3,4-Dimethoxyphenylboronic acid Pinacol ester, 1,2-Dimethoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene, 2-(3,4-Dimethoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

021828-5G
FLUO021828-5GEA 137 EUR
FLUO021828-5G
3,4-Dimethoxyphenylboronsäurepinakolester
3,4-Dimethoxyphenylboronsäurepinakolester
Formel: C₁₄H₂₁BO₄
Molecular Weight: 264.13 g/mol
MDL: MFCD05863909
CAS-Nummer: 365564-10-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR