2-Methoxyphenylboronsäurepinakolester

Lieferant: FLUOROCHEM

Synonyme: 2-Methoxybenzeneboronic acid pinacol ester, 2-Methoxyphenylboronic acid, pinacol ester, 2-(2-Methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

021829-5G
FLUO021829-5GEA 177 EUR
FLUO021829-5G
2-Methoxyphenylboronsäurepinakolester
2-Methoxyphenylboronsäurepinakolester
Formel: C₁₃H₁₉BO₃
Molecular Weight: 234,10 g/mol
MDL: MFCD05863916
CAS-Nummer: 190788-60-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR