4-Biphenylboronsäure

Lieferant: FLUOROCHEM

Synonyme: [1,1'-Biphenyl]-4-ylboronic acid

021833-25G 021833.5G
FLUO021833-25GEA 128 EUR
FLUO021833-25G FLUO021833.5G
4-Biphenylboronsäure
4-Biphenylboronsäure
Formel: C₁₂H₁₁BO₂
Molecular Weight: 198,03 g/mol
MDL: MFCD00093311
CAS-Nummer: 5122-94-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR