4-(Trifluormethyl)phenylboronsäurepinakolester

Lieferant: FLUOROCHEM

Synonyme: 4,4,5,5-Tetramethyl-2-(4-(trifluoromethyl)phenyl)-1,3,2-dioxaborolane, 4-Trifluoromethylphenylboronic acid pinacol ester

021836-5G
FLUO021836-5GEA 272 EUR
FLUO021836-5G
4-(Trifluormethyl)phenylboronsäurepinakolester
4-(Trifluormethyl)phenylboronsäurepinakolester
Formel: C₁₃H₁₆BF₃O₂
Molecular Weight: 272,0712 g/mol
MDL: MFCD05863924
CAS-Nummer: 214360-65-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR