3-Acetylphenylboronsäurepinakolester
Lieferant: FLUOROCHEM
Synonyme:
1-(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)ethanone, 3-Acetylphenylboronic acid, pinacol ester
021838-5G
FLUO021838-5GEA
354
EUR
FLUO021838-5G
3-Acetylphenylboronsäurepinakolester
3-Acetylphenylboronsäurepinakolester
Formel:
C₁₄H₁₉BO₃ Molecular Weight: 246,1099 g/mol |
MDL:
MFCD05863923 CAS-Nummer: 214360-49-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...