3-(Trifluormethoxy)phenylboronsäurepinakolester

Lieferant: FLUOROCHEM

Synonyme: 3-(Trifluoromethoxy)phenylboronic acid Pinacol ester, 4,4,5,5-Tetramethyl-2-[3-(trifluoromethoxy)phenyl]-1,3,2-dioxaborolane, 1-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-3-(trifluoromethoxy)benzene

021842-5G
FLUO021842-5GEA 515 EUR
FLUO021842-5G
3-(Trifluormethoxy)phenylboronsäurepinakolester
3-(Trifluormethoxy)phenylboronsäurepinakolester
Formel: C₁₃H₁₆BF₃O₃
Molecular Weight: 288.0706 g/mol
MDL: MFCD05863919
CAS-Nummer: 262376-31-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR