2-(Trifluormethoxy)phenylboronsäurepinakolester

Lieferant: FLUOROCHEM

Synonyme: 4,4,5,5-Tetramethyl-2-(2-(trifluoromethoxy)phenyl)-1,3,2-dioxaborolane

021843-5G
FLUO021843-5GEA 650 EUR
FLUO021843-5G
2-(Trifluormethoxy)phenylboronsäurepinakolester
2-(Trifluormethoxy)phenylboronsäurepinakolester
Formel: C₁₃H₁₆BF₃O₃
Molecular Weight: 288,0706 g/mol
MDL: MFCD05863918
CAS-Nummer: 832114-04-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR