3-Cyan-4-fluorphenylboronsäure

Lieferant: FLUOROCHEM

Synonyme: 3-Cyano-4-fluorobenzeneboronic acid

021844-5G
FLUO021844-5GEA 209 EUR
FLUO021844-5G
3-Cyan-4-fluorphenylboronsäure
3-Cyan-4-fluorphenylboronsäure
Formel: C₇H₅BFNO₂
Molecular Weight: 164,93 g/mol
MDL: MFCD03095130
CAS-Nummer: 214210-21-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR