4-Fluor-2-methylphenylboronsäure

Lieferant: FLUOROCHEM

Synonyme: 4-Fluoro-2-methylbenzeneboronic acid, 2-Methyl-4-fluorophenylboronic acid

021849-5G
FLUO021849-5GEA 132 EUR
FLUO021849-5G
4-Fluor-2-methylphenylboronsäure
4-Fluor-2-methylphenylboronsäure
Formel: C₇H₈BFO₂
Molecular Weight: 153,95 g/mol
MDL: MFCD02093072
CAS-Nummer: 139911-29-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR