4-(4-Methyl-1-piperazinylcarbonyl)phenylboronsäurepinakolester

Lieferant: FLUOROCHEM

Synonyme: 4-(4-Methyl-1-piperazinylcarbonyl)benzeneboronic acid pinacol ester, 4-(4-Methylpiperazine-1-carbonyl)phenylboronic acid, pinacol ester, (4-Methylpiperazin-1-yl)(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanone

021850-5G
FLUO021850-5GEA 513 EUR
FLUO021850-5G
4-(4-Methyl-1-piperazinylcarbonyl)phenylboronsäurepinakolester
4-(4-Methyl-1-piperazinylcarbonyl)phenylboronsäurepinakolester
Formel: C₁₈H₂₇BN₂O₃
Molecular Weight: 330,23 g/mol
MDL: MFCD05863914
CAS-Nummer: 832114-06-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR