4-(2-(Dimethylamino)ethylcarbamoyl)phenylboronsäurepinakolester

Lieferant: FLUOROCHEM

Synonyme: N-(2-Dimethylaminoethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide, 4-(N-[2-(Dimethylamino)ethyl]carbamoyl)benzeneboronic acid pinacol ester

021857-5G
FLUO021857-5GEA 459 EUR
FLUO021857-5G
4-(2-(Dimethylamino)ethylcarbamoyl)phenylboronsäurepinakolester
4-(2-(Dimethylamino)ethylcarbamoyl)phenylboronsäurepinakolester
Formel: C₁₇H₂₇BN₂O₃
MDL: MFCD05864307
CAS-Nummer: 832114-11-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR