3-Brom-4-(trifluormethoxy)phenol

Lieferant: FLUOROCHEM

Synonyme: 1-Brom-3-hydroxy-6-(trifluormethoxy)benzol

021863-5G
FLUO021863-5GEA 139 EUR
FLUO021863-5G
3-Brom-4-(trifluormethoxy)phenol
3-Brom-4-(trifluormethoxy)phenol
Formel: C₇H₄BrF₃O₂
MDL: MFCD05864310
CAS-Nummer: 886496-88-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR