1-Brom-2-fluor-4-nitrobenzol

Lieferant: FLUOROCHEM

Synonyme: 4-Brom-3-fluornitrobenzol

021894-5G 021894-25G
FLUO021894-5GEA 34.1 EUR
FLUO021894-5G FLUO021894-25G
1-Brom-2-fluor-4-nitrobenzol
1-Bromo-2-fluoro-4-nitrobenzene
Formel: C₆H₃BrFNO₂
Molecular Weight: 220 g/mol
MDL: MFCD05865088
CAS-Nummer: 185331-69-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR