2,4-Dichlor-6,7-dimethoxychinazolin

Lieferant: FLUOROCHEM

021895-1G
FLUO021895-1GEA 57.8 EUR
FLUO021895-1G
2,4-Dichlor-6,7-dimethoxychinazolin
2,4-Dichloro-6,7-dimethoxyquinazoline
Formel: C₁₀H₈Cl₂N₂O₂
Molecular Weight: 259,0887 g/mol
MDL: MFCD00051733
CAS-Nummer: 27631-29-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR