2-Formyl-6-picolin

Lieferant: FLUOROCHEM

021900-5G 021900-25G
FLUO021900-5GEA 84.9 EUR
FLUO021900-5G FLUO021900-25G
2-Formyl-6-picolin
2-Formyl-6-picolin
Formel: C₇H₇NO
MDL: MFCD00006291
CAS-Nummer: 1122-72-1
EINECS: 214-359-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR