3-Fluor-5-nitrobenzonitril

Lieferant: FLUOROCHEM

022023-1G
FLUO022023-1GEA 180 EUR
FLUO022023-1G
3-Fluor-5-nitrobenzonitril
3-Fluor-5-nitrobenzonitril
Formel: C₇H₃FN₂O₂
Molecular Weight: 166,11 g/mol
MDL: MFCD05864356
CAS-Nummer: 110882-60-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR