3-Cyan-5-fluoranilin

Lieferant: FLUOROCHEM

Synonyme: 5-Amino-3-fluorbenzonitril, 5-Amino-3-fluorbenzoesäurenitril

022024-1G
FLUO022024-1GEA 0 EUR
FLUO022024-1G
3-Cyan-5-fluoranilin
3-Cyan-5-fluoranilin
Formel: C₇H₅FN₂
Molecular Weight: 136,13 g/mol
MDL: MFCD05864357
CAS-Nummer: 210992-28-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR