3-Fluor-5-nitrobenzoesäure

Lieferant: FLUOROCHEM

022027-1G
FLUO022027-1GEA 67.1 EUR
FLUO022027-1G
3-Fluor-5-nitrobenzoesäure
3-Fluor-5-nitrobenzoesäure
Formel: C₇H₄FNO₄
MDL: MFCD05864360
CAS-Nummer: 14027-75-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR