2'-Hydroxy-5'-methyl-2,2,2-trifluoracetophenon

Lieferant: FLUOROCHEM

Synonyme: 2,2,2-trifluoro-1-(2-hydroxy-5-methylphenyl)ethanone

022052-1G
FLUO022052-1GEA 645 EUR
FLUO022052-1G
2'-Hydroxy-5'-methyl-2,2,2-trifluoracetophenon
2'-Hydroxy-5'-methyl-2,2,2-trifluoracetophenon
Formel: C₉H₇F₃O₂
MDL: MFCD05864383
CAS-Nummer: 70978-57-3
EINECS: 275-078-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR