4'-Methoxy-2,2,2-trifluoracetophenon

Lieferant: FLUOROCHEM

Synonyme: 2,2,2-Trifluoro-1-(4-methoxyphenyl)ethanone

022054-1G
FLUO022054-1GEA 128 EUR
FLUO022054-1G
4'-Methoxy-2,2,2-trifluoracetophenon
4'-Methoxy-2,2,2-trifluoracetophenon
Formel: C₉H₇F₃O₂
Molecular Weight: 204.1500 g/mol
MDL: MFCD00018061
CAS-Nummer: 711-38-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR