2-Amino-4,4,4-trifluorbuttersäure Hydrochlorid

Lieferant: FLUOROCHEM

Synonyme: (±)-2-Amino-4,4,4-trifluorbuttersäure Hydrochlorid

022071-1G
FLUO022071-1GEA 0 EUR
FLUO022071-1G
2-Amino-4,4,4-trifluorbuttersäure Hydrochlorid
2-Amino-4,4,4-trifluorbuttersäure Hydrochlorid
Formel: C₄H₇ClF₃NO₂
MDL: MFCD00671488
CAS-Nummer: 15959-93-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR