trans-2-Phenylcyclohexanol

Lieferant: FLUOROCHEM

022164-1G
FLUO022164-1GEA 180 EUR
FLUO022164-1G
trans-2-Phenylcyclohexanol
trans-2-Phenylcyclohexanol
Formel: C₁₂H₁₆O
MDL: MFCD00003865
CAS-Nummer: 2362-61-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5b6044e2d2d4bf696fab754466269933e6b9afae&vid=5b6044e2d2d4bf696fab754466269933e6b9afae&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR