2-(4-tert-Butylphenyl)ethanol
Lieferant: FLUOROCHEM
Synonyme:
2-(4-(tert-Butyl)phenyl)ethanol
022224-1G
FLUO022224-1GEA
30.8
EUR
FLUO022224-1G
2-(4-tert-Butylphenyl)ethanol
2-(4-tert-Butylphenyl)ethanol
Formel:
C₁₂H₁₈O Molecular Weight: 178,28 g/mol |
MDL:
MFCD00236023 CAS-Nummer: 5406-86-0 EINECS: 410-020-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...