4-(3-Chlor-5-fluorphenyl)-1-buten-4-ol
Lieferant: FLUOROCHEM
022226-1G
FLUO022226-1GEA
1160
EUR
FLUO022226-1G
4-(3-Chlor-5-fluorphenyl)-1-buten-4-ol
4-(3-Chlor-5-fluorphenyl)-1-buten-4-ol
Formel:
C₁₀H₁₀ClFO |
MDL:
MFCD06201122 CAS-Nummer: 842123-72-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...