4-(4-Chlor-2-methylphenyl)-1-buten-4-ol

Lieferant: FLUOROCHEM

022227-1G
FLUO022227-1GEA 1160 EUR
FLUO022227-1G
4-(4-Chlor-2-methylphenyl)-1-buten-4-ol
4-(4-Chlor-2-methylphenyl)-1-buten-4-ol
Formel: C₁₁H₁₃ClO
MDL: MFCD06201123
CAS-Nummer: 842123-73-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR