4-(4-Chlor-3-methylphenyl)-1-buten-4-ol

Lieferant: FLUOROCHEM

022228-1G
FLUO022228-1GEA 1160 EUR
FLUO022228-1G
4-(4-Chlor-3-methylphenyl)-1-buten-4-ol
4-(4-Chlor-3-methylphenyl)-1-buten-4-ol
Formel: C₁₁H₁₃ClO
MDL: MFCD06201124
CAS-Nummer: 842123-74-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR