4-(3,5-Dichlorphenyl)-1-buten-4-ol

Lieferant: FLUOROCHEM

022229-1G
FLUO022229-1GEA 1160 EUR
FLUO022229-1G
4-(3,5-Dichlorphenyl)-1-buten-4-ol
4-(3,5-Dichlorphenyl)-1-buten-4-ol
Formel: C₁₀H₁₀Cl₂O
MDL: MFCD06201125
CAS-Nummer: 842123-75-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR