4-(3,4-Dimethoxyphenyl)-1-buten-4-ol

Lieferant: FLUOROCHEM

022230-1G
FLUO022230-1GEA 1170 EUR
FLUO022230-1G
4-(3,4-Dimethoxyphenyl)-1-buten-4-ol
4-(3,4-Dimethoxyphenyl)-1-buten-4-ol
Formel: C₁₂H₁₆O₃
MDL: MFCD00598697
CAS-Nummer: 5452-46-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR