4-(3,4-Dimethylphenyl)-1-buten-4-ol
Lieferant: FLUOROCHEM
022232-1G
FLUO022232-1GEA
1160
EUR
FLUO022232-1G
4-(3,4-Dimethylphenyl)-1-buten-4-ol
4-(3,4-Dimethylphenyl)-1-buten-4-ol
Formel:
C₁₂H₁₆O |
MDL:
MFCD06201127 CAS-Nummer: 842123-77-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...