4-(3-Fluor-4-methylphenyl)-1-buten-4-ol

Lieferant: FLUOROCHEM

022238-1G
FLUO022238-1GEA 1160 EUR
FLUO022238-1G
4-(3-Fluor-4-methylphenyl)-1-buten-4-ol
4-(3-Fluor-4-methylphenyl)-1-buten-4-ol
Formel: C₁₁H₁₃FO
MDL: MFCD06201133
CAS-Nummer: 842123-82-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR