4-(4-Fluor-3-methylphenyl)-1-buten-4-ol
Lieferant: FLUOROCHEM
022239-1G
FLUO022239-1GEA
1160
EUR
FLUO022239-1G
4-(4-Fluor-3-methylphenyl)-1-buten-4-ol
4-(4-Fluor-3-methylphenyl)-1-buten-4-ol
Formel:
C₁₁H₁₃FO |
MDL:
MFCD06201134 CAS-Nummer: 842123-83-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...