4-(3-Bromphenyl)-1-buten-4-ol

Lieferant: FLUOROCHEM

022241-1G
FLUO022241-1GEA 0 EUR
FLUO022241-1G
4-(3-Bromphenyl)-1-buten-4-ol
4-(3-Bromphenyl)-1-buten-4-ol
Formel: C₁₀H₁₁BrO
MDL: MFCD06201136
CAS-Nummer: 114095-73-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR