4-(4-Bromphenyl)-1-buten-4-ol

Lieferant: FLUOROCHEM

022242-1G
FLUO022242-1GEA 1160 EUR
FLUO022242-1G
4-(4-Bromphenyl)-1-buten-4-ol
4-(4-Bromphenyl)-1-buten-4-ol
Formel: C₁₀H₁₁BrO
MDL: MFCD06201137
CAS-Nummer: 113842-31-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR