4-(3-Chlorphenyl)-1-buten-4-ol

Lieferant: FLUOROCHEM

022243-1G
FLUO022243-1GEA 932 EUR
FLUO022243-1G
4-(3-Chlorphenyl)-1-buten-4-ol
4-(3-Chlorphenyl)-1-buten-4-ol
Formel: C₁₀H₁₁ClO
MDL: MFCD06201138
CAS-Nummer: 67472-21-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR