4-(4-Chlorphenyl)-1-buten-4-ol

Lieferant: FLUOROCHEM

Synonyme: 1-(4-Chlorophenyl)-3-buten-1-ol, 1-(4-Chlorophenyl)but-3-en-1-ol

022244-1G
FLUO022244-1GEA 0 EUR
FLUO022244-1G
4-(4-Chlorphenyl)-1-buten-4-ol
4-(4-Chlorphenyl)-1-buten-4-ol
Formel: C₁₀H₁₁ClO
Molecular Weight: 182.65 g/mol
MDL: MFCD03427246
CAS-Nummer: 14506-33-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR