3-Chlor-5-fluorbenzylalkohol

Lieferant: FLUOROCHEM

022246-1G
FLUO022246-1GEA 49.1 EUR
FLUO022246-1G
3-Chlor-5-fluorbenzylalkohol
3-Chlor-5-fluorbenzylalkohol
Formel: C₇H₆ClFO
Molecular Weight: 160,58 g/mol
MDL: MFCD03788554
CAS-Nummer: 79944-64-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR