4-Chlor-2-methylbenzylalkohol

Lieferant: FLUOROCHEM

022248-1G
FLUO022248-1GEA 119 EUR
FLUO022248-1G
4-Chlor-2-methylbenzylalkohol
4-Chlor-2-methylbenzylalkohol
Formel: C₈H₉ClO
Molecular Weight: 156,61 g/mol
MDL: MFCD06201140
CAS-Nummer: 129716-11-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR