4-Chlor-3-methylbenzylalkohol

Lieferant: FLUOROCHEM

022249-1G
FLUO022249-1GEA 1110 EUR
FLUO022249-1G
4-Chlor-3-methylbenzylalkohol
4-Chlor-3-methylbenzylalkohol
Formel: C₈H₉ClO
Molecular Weight: 156,61 g/mol
MDL: MFCD06201141
CAS-Nummer: 131271-19-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR