3-(1,3-Dioxolan-2-yl)propiophenon

Lieferant: FLUOROCHEM

Synonyme: 3-(1,3-Dioxolan-2-yl)-1-phenylpropan-1-one

022277-1G
FLUO022277-1GEA 0 EUR
FLUO022277-1G
3-(1,3-Dioxolan-2-yl)propiophenon
3-(1,3-Dioxolan-2-yl)propiophenon
Formel: C₁₂H₁₄O₃
Molecular Weight: 206,237 g/mol
MDL: MFCD06201162
CAS-Nummer: 54743-42-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR