2-(1,3-Dioxolan-2-ylmethyl)anisol

Lieferant: FLUOROCHEM

022279-1G
FLUO022279-1GEA 794 EUR
FLUO022279-1G
2-(1,3-Dioxolan-2-ylmethyl)anisol
2-(1,3-Dioxolan-2-ylmethyl)anisol
Formel: C₁₁H₁₄O₃
MDL: MFCD04117674
CAS-Nummer: 842123-92-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR