3-(1,3-Dioxolan-2-ylmethyl)anisol

Lieferant: FLUOROCHEM

022282-1G
FLUO022282-1GEA 794 EUR
FLUO022282-1G
3-(1,3-Dioxolan-2-ylmethyl)anisol
3-(1,3-Dioxolan-2-ylmethyl)anisol
Formel: C₁₁H₁₄O₃
MDL: MFCD04117675
CAS-Nummer: 842123-95-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR