4'-Brom-3-(1,3-dioxolan-2-yl)propiophenon
Lieferant: FLUOROCHEM
022283-1G
FLUO022283-1GEA
0
EUR
FLUO022283-1G
4'-Brom-3-(1,3-dioxolan-2-yl)propiophenon
4'-Brom-3-(1,3-dioxolan-2-yl)propiophenon
Formel:
C₁₂H₁₃BrO₃ |
MDL:
MFCD06201163 CAS-Nummer: 842123-96-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...