3'-Brom-3-(1,3-dioxolan-2-yl)propiophenon
Lieferant: FLUOROCHEM
022284-1G
FLUO022284-1GEA
0
EUR
FLUO022284-1G
3'-Brom-3-(1,3-dioxolan-2-yl)propiophenon
3'-Brom-3-(1,3-dioxolan-2-yl)propiophenon
Formel:
C₁₂H₁₃BrO₃ |
MDL:
MFCD06201164 CAS-Nummer: 842123-97-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...