4-(1,3-Dioxolan-2-ylmethyl)anisol

Lieferant: FLUOROCHEM

022288-1G
FLUO022288-1GEA 794 EUR
FLUO022288-1G
4-(1,3-Dioxolan-2-ylmethyl)anisol
4-(1,3-Dioxolan-2-ylmethyl)anisol
Formel: C₁₁H₁₄O₃
MDL: MFCD04117676
CAS-Nummer: 91970-78-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR